Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure
Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure
Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure
Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure
Thermo Scientific™

Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure

Store corrosive chemicals and distilled water with these large, narrow-mouth PPCO bottles, which are resistant to most acids, bases and alcohols.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeUnit Size
2203-00052 L38-430 mmCase of 6
2203-0005PK
also known as 2203-0005
2 L38-430 mmEach
2203-00104 L38-430 mmCase of 6
2203-0010PK
also known as 2203-0010
4 L38-430 mmEach
2203-00208 L53BCase of 6
2203-0020PK
also known as 2203-0020
8 L53BEach
Catalog number 2203-0005
Price (USD)
148.49
Online exclusive
212.14
Save 63.65
Case of 6
Capacity (Metric):
2 L
Closure Size:
38-430 mm
Unit Size:
Case of 6
Price (USD)
148.49
Online exclusive
212.14
Save 63.65
Case of 6

Store corrosive chemicals and distilled water with Thermo Scientific™ Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure, which are resistant to most acids, bases and alcohols. Large sizes of these polypropylene copolymer bottles have a built-in shoulder loop.

Features:
  • 4 and 8L feature a built-in shoulder loop on larger sizes for attaching identification tag
  • Lighter weight than glass alternatives make for easy lifting
  • Translucence allows for easy viewing of liquid levels
  • Linerless caps ensure leakproof† protection without the use of liner that can wrinkle, cause leaks, or contaiminate your reagents
  • Autoclavable/Leakproof† when used with Nalgene closures
  • Manufactured from non-toxic materials

Includes:

Polypropylene screw caps

Note:

Completely disengage threads or remove closure before autoclaving. Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  These bottles and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene PP and PPCO bottles if services are available in your community.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
AutoclavableAutoclavable
Capacity0.5 gal., 2 L
Capacity (English)0.5 gal.
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure ColorWhite
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size38-430 mm
Closure TypeScrew Cap
DescriptionLarge Narrow-Mouth Bottle PP 2L
Diameter (Metric)121 mm
Diameter (Metric) Inner Neck25 mm
DisposableReusable
Height (English)9.8 in.
Height (Metric)249 mm
Neck Finish38 mm
No. per Case6
No. per Pack1
Product TypeNarrow Mouth Bottle
ShapeRound
Through the Closure DispensingNo
Width (English)4.75 in.
Capacity (Metric)2 L
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialPPCO
MouthNarrow
SterilityNon-sterile
Unit SizeCase of 6

Frequently asked questions (FAQs)

Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.

What do the terms IBM and EBM refer to on the Certificate of Conformity (COC) for plastic consumables?

Please see the definitions for each term below:

  • IBM: injection blow molded
  • EBM: extrusion blow molded