Fiberlite 500mL Bottle, Polypropylene (PPCO) - FAQs
Fiberlite 500mL Bottle, Polypropylene (PPCO) - FAQs
View additional product information for Fiberlite 500mL Bottle - FAQs (010-1493, 010-1494)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.