dNTP Set (100 mM), 8 x 1.25 mL - FAQs
View additional product information for dNTP Set (100 mM) - FAQs (10297117, 10297018)
2 product FAQs found
Why do some dNTP mixes contain dTTP and others TTP?
Thymine is normally found as a deoxyribose. Thus, TTP and dTTP are essentially the same thing. Both types of mixes can be used in PCR reactions
What is the final concentration of each deoxynucleotide in your 100 mM dNTP set (Cat. Nos. 10297018, 10297117)?
Each deoxynucleotide (dATP, dCTP, dGTP, dTTP) is supplied at a concentration of 100 mM.