Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • F4/80 Antibodies

          Invitrogen

          F4/80 Monoclonal Antibody (BM8), FITC, eBioscience™

          98 Published Figures
          452 References
          View all (77) F4/80 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite F4/80 Monoclonal Antibody (BM8), FITC, eBioscience™

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (98)
          F4/80 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          F4/80 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 99

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          F4/80 Antibody (11-4801-81) in Flow

          C57BL/6 mouse resident peritoneal macrophages were stained with VSIG4 Monoclonal Antibody, APC (Product # 17-5752-82) and 0.5 µg of Rat IgG2a kappa Isotype Control, FITC (Product # 11-4321-81) (left) or 0.5 µg of F4/80 Monoclonal Antibody, FITC (right). Viable cells in the large scatter population were used for analysis, as determined by 7-AAD (Product # 00-6993-50). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          F4/80 Antibody in Immunohistochemistry, Immunocytochemistry, Flow Cytometry (IHC, ICC/IF, Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          F4/80 Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)
          F4/80 Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          11-4801-81

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 94 publications 94 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 5 publications 5 publications

          Immunocytochemistry (ICC/IF)

          -
          View 26 publications 26 publications

          Flow Cytometry (Flow)

          0.5 µg/test
          View 321 publications 321 publications

          Functional Assay (Functional)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Hamster, Human, Mouse, Rat

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), FITC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          BM8

          Conjugate

          FITC FITC FITC
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 532 
          • Alexa Fluor 561
          • Alexa Fluor 660 
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 563
          • Brilliant UV 661
          • Brilliant UV 805
          • Brilliant Violet 421
          • Brilliant Violet 650
          • Brilliant Violet 711
          • eFluor 450
          • eFluor 506
          • eFluor 570
          • eFluor 660
          • Functional Grade
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 702
          • Super Bright 780
          • Request custom conjugation

          Excitation/Emission Max

          498/517 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2735037

          Product Specific Information

          Description: The BM8 monoclonal antibody reacts with mouse F4/80 antigen, an approximately 160 kDa surface receptor. It belongs to the EGF-TM7 family of proteins. As such it contains seven EGF-like domains on its extracellular N-terminus, seven transmembrane spanning sequences, and an intracellular C-terminal domain showing homology to other TM7 superfamily members. The F4/80 antigen is expressed by a majority of mature macrophages and is one of the best markers for this population of cells. However, other cell types, such as peritoneal eosinophils, Langerhans cells, and some other dendritic cell subtypes, have been reported to express this antigen as well. Expression of F4/80 commences during early myeloid development in vivo and can be upregulated on BM cells stimulated in vitro with M-CSF. Some populations of macrophages, especially in the lymphoid microenvironment, may be devoid of F4/80.

          Applications Reported: BM8 has been reported for use in flow cytometric analysis.

          Applications Tested: The BM8 antibody has been tested by flow cytometric analysis of mouse resident peritoneal exudate cells. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          F4/80 antigen is a 160 kDa cell surface glycoprotein that is a member of the EGF TM7 family of proteins and shares 68% overall amino acid identity with human EGF module containing mucin like hormone receptor 1 (EMR1). Expression of F4/80 is heterogeneous and is reported to vary during macrophage maturation and activation. The F4/80 antigen is expressed on a wide range of mature tissue macrophages including Kupffer cells, Langerhans, microglia, macrophages located in the gut lamina propria, peritoneal cavity, lung, thymus, bone marrow stroma and macrophages in the red pulp of the spleen. F4/80 expression has also been reported on a subpopulation of dendritic cells but is absent from macrophages located in T cell areas of the spleen and lymph node. The ligands and biological functions of the F4/80 antigen have not yet been determined but recent studies suggest a role for F4/80 in the generation of efferent CD8+ve regulatory T cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: cell surface glycoprotein F4/80; EGF-like module containing, mucin-like, hormone receptor-like sequence 1; EGF-like module receptor 1; EGF-like module-containing mucin-like hormone receptor-like 1; EMR1 hormone receptor; unnamed protein product

          View more View less

          Gene Aliases: DD7A5-7; EGF-TM7; Emr1; F4/80; Gpf480; Ly71; TM7LN3

          View more View less

          UniProt ID: (Mouse) Q61549

          View more View less

          Entrez Gene ID: (Mouse) 13733

          View more View less

          Function(s)
          transmembrane signaling receptor activity G-protein coupled receptor activity calcium ion binding
          Process(es)
          cell surface receptor signaling pathway G-protein coupled receptor signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-p95z8:80/100.66.74.151:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline