Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MSTATTVAP AGIPATPGPV NPPPPEVSNP SKPGRKTNQL QYMQNVVVKT LWKHQFAWPF YQPVDAIKLN LPDYHKIIKN PMDMGTIKKR LENNYYWSAS ECMQDFNTMF TNCYIYNKPT DDIVLMAQAL EKIFLQKVAQ MPQEEVELLP PAPKGKGRKP AAGAQSAGTQ QVAAVSSVSP ATPFQSVPPT VSQTPVIAAT PVPTITANVT SVPVPPAAAP PPPATPIVPV VPPTPPVVKK KGVKRKADTT TPTTSAITAS RSESPPPLSD PKQAKVVARR ESGGRPIKPP KKDLEDGEVP QHAGKKGKLS EHLRYCDSIL REMLSKKHAA YAWPFYKPVD AEALELHDYH DIIKHPMDLS TVKRKMDGRE YPDAQGFAAD VRLMFSNCYK YNPPDHEVVA M (1-400 aa encoded by BC032124)
This gene was identified based on its homology to the gene encoding the RING3 protein, a serine/threonine kinase. The gene localizes to 9q34, a region which contains several major histocompatibility complex (MHC) genes. The function of the encoded protein is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bromodomain-containing protein 3; female sterile homeotic related gene 2; KIAA0043; mitotic chromatin binding capacity; reprogramming promoting activity; RING3-like protein; unnamed protein product
Gene Aliases: BRD3; FSHRG2; KIAA0043; ORFX; RING3L
UniProt ID: (Human) Q15059
Entrez Gene ID: (Human) 8019
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support