B-27™ Supplement (50X), minus vitamin A, 10 mL - FAQs
B-27™ Supplement (50X), minus vitamin A, 10 mL - FAQs
View additional product information for B-27™ Supplement (50X), minus vitamin A - FAQs (12587010, 12587001)
8 product FAQs found
Can B-27 supplement be filtered?
Yes, the protein content in this product is high enough so filtering through a low protein binding filter as a 50X or 1X in solution should not be a problem.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can Neurobasal Medium alone or Neurobasal Medium supplemented with B-27 supplement be frozen to extend product shelf life?
No, it is not recommended to freeze media due to the potential of precipitates forming upon thaw. Inorganic salts and amino acids in the formulation may come out of solution when exposed to temperature fluctuations. These precipitates will not go back into solution easily once formed.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What supplements should I use with Neurobasal or Neurobasal-A medium?
Neurobasal and Neurobasal-A media (for postnatal and adult neurons) allow for long-term maintenance of neuronal cells without the need for an astrocyte feeder layer. These media should be supplemented with either serum or a serum-free supplement, plus 0.5mM L-glutamine. B-27 supplement is a serum-free supplement that comes as a 50X concentrate in a 10ml volume. This is enough supplement for 500ml of media. Fetal, postnatal, and adult neural cultures can be grown in the appropriate Neurobasal medium supplemented with B-27 supplement .
We also have two other supplements. One is called G-5 and is for growth and expression of glial cells (normal and tumor) of astrocytic phenotype. This comes in a 1ml size, at a 100X concentration. The other supplement is called N-2 and is for growth and expression of post-mitotic neurons and tumor cells of neuronal phenotype. This comes in a 5ml size, at a 100X concentration.
For more information on Neurobasal media, search "Neurobasal" from our website home page.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Do you offer B-27 supplements with and without insulin?
Yes, we do offer B-27 Supplement, minus insulin (Cat. Nos. A1895601 and A1895602). All our other B-27 supplements do contain insulin.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I thaw B-27 Supplements in a 37 degrees C water bath or incubator?
We do not recommend thawing at 37 degrees C. Best results are observed upon thawing at room temperature for approximately 2 hrs or overnight at 2-8 degrees C.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How do I thaw B-27 Supplements?
We recommend thawing it at 4 degrees C overnight or at room temperature for 2 hours. We don't recommend thawing in a water bath or incubator.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I re-freeze B-27 Supplements into smaller aliquots for future use?
Yes, we recommend that you aliquot when you first thaw it and thereafter subject the aliquot to no more than 2 additional freeze/thaw cycles.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What B-27 supplements do you offer?
We offer the following B-27 supplement products, which are developed for specific applications:
- Gibco B-27 Serum-free Supplement (Cat. No. 17504044): Complete formulation for the growth of long-term viability of neurons.
- Gibco B-27 Supplement minus AO (antioxidants) (Cat. No. 10889038): Five antioxidants are removed. The supplement is ideal in studies of oxidative damage, apoptosis, or applications in which free radical damage to neurons occurs.
- Gibco B-27 Supplement minus Insulin (Cat. No. A1895601): Insulin is removed. The supplement is ideal for studies of insulin secretion and insulin receptors.
- Gibco B-27 Supplement minus Vitamin A (Cat. No. 12587010): Vitamin A which induces neural differentiation is removed. The supplement is ideal for proliferation of stem cells.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.