Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAEERVATR TQFPVSTESQ KPRQKKAPEF PILEKQNWLI HLHYIRKDYE ACKAVIKEQL QETQGLCEYA IYVQALIFRL EGNIQESLEL FQTCAVLSPQ SADNLKQVAR SLFLLGKHKA AIEVYNEAAK LNQKDWEISH NLGVCYIYLK QFNKAQDQLH NALNLNRHDL TYIMLGKIHL LEGDLDKAIE VYKKAVEFSP ENTELLTTLG LLYLQLGIYQ KAFEHLGNAL TYDPTNYKAI LAAGSMMQTH GDFDVALTKY RVVACAVPES PPLWNNIGMC FFGKKKYVAA ISCLKRANYL APFDWKILYN LGLVHLTMQQ YASAFHFLSA AINFQPKMGE LYMLLAVALT NL (1-351 aa encoded by BC027624)
BBS4 contains tetratricopeptide repeats (TPR), similar to O-linked N-acetyglucosamine transferase. Mutations in the gene encoding this protein have been observed in patients with Bardet-Biedl syndrome type 4. BBS4 may play a role in pigmentary retinopathy, obesity, polydactyly, renal malformation and mental retardation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bardet-Biedl syndrome 4 homolog; Bardet-Biedl syndrome 4 protein; Bardet-Biedl syndrome 4 protein homolog; BBS4; BBSome complex member BBS4; unnamed protein product
Gene Aliases: AW537059; AW742241; BBS4; D9Ertd464e
UniProt ID: (Human) Q96RK4, (Mouse) Q8C1Z7
Entrez Gene ID: (Human) 585, (Mouse) 102774, (Rat) 300754
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support