CXCL12 alpha (SDF-1 alpha) Polyclonal Antibody, eBioscience™ - FAQs
CXCL12 alpha (SDF-1 alpha) Polyclonal Antibody, eBioscience™ - FAQs
View additional product information for CXCL12 alpha (SDF-1 alpha) Polyclonal Antibody, eBioscience™ - FAQs (14-7992-83, 14-7992-81)
1 product FAQs found
Can we use this antibody, CXCL12 alpha (SDF-1 alpha) Polyclonal Antibody, eBioscience, with human, mouse, and rat tissue?
This antibody is reactive towards mouse and rat and not human samples.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.