Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Description: This 31B monoclonal antibody reacts with human CD208, which is also known as dendritic cell lysosome-associated membrane glycoprotein (DC-LAMP). CD208 is expressed almost exclusively by mature dendritic cells. Expression of this molecule can be induced in monocytes by stimulation with GM-CSF and IL-4. Moreover, CD208 can be upregulated in CD34+-derived dendritic cells by GM-CSF, SCF, and TNFa. CD208 is also expressed in dendritic cells of non-lymphoid tissues such as skin. While CD208 can be detected on the cell surface, this protein is mainly localized within lysosomes. CD208 plays a key role in the processing and presentation of antigen during the immune response.
The 31B antibody only recognizes intracellular CD208, not the cell surface-expressed protein.
Applications Reported: This 31B antibody has been reported for use in intracellular staining followed by flow cytometric analysis.
Applications Tested: This 31B antibody has been pre-titrated and tested by flow cytometric analysis of in vitro differentiated mature dendritic cells. This can be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.
Filtration: 0.2 µm post-manufacturing filtered.
Dendritic cells (DCs) are the most potent antigen-presenting cells. Immature DCs efficiently capture antigens and differentiate into interdigitating dendritic cells (IDCs) in lymphoid tissues that induce primary T-cell responses (summary by de Saint-Vis et al., 1998 [PubMed 9768752]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: DC-lysosome-associated membrane glycoprotein; lysosomal associated membrane protein 3; unnamed protein product
Gene Aliases: CD208; DC LAMP; DC-LAMP; DCLAMP; LAMP; LAMP-3; TSC403
UniProt ID: (Human) Q9UQV4
Entrez Gene ID: (Human) 27074
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support