Can exo-Klenow be purchased separately from your array labeling kits?
Yes, the catalog number for the exo-Klenow is AM2008. The enzyme concentration is 5 U/µL.
Search
View additional product information for BioPrime™ DNA Labeling System - FAQs (18094011)
1 product FAQs found