Nalgene™ Natural PPCO Diagnostic Bottles without Closure: Bulk Pack, 3.4L - FAQs
View additional product information for Nalgene™ Natural PPCO Diagnostic Bottles without Closure: Bulk Pack - FAQs (362006-9050, 362006-9125, 362006-9025)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.