Nalgene™ Natural PPCO Diagnostic Bottles without Closure: Bulk Pack
Nalgene™ Natural PPCO Diagnostic Bottles without Closure: Bulk Pack
Nalgene™ Natural PPCO Diagnostic Bottles without Closure: Bulk Pack
Nalgene™ Natural PPCO Diagnostic Bottles without Closure: Bulk Pack
Thermo Scientific™

Nalgene™ Natural PPCO Diagnostic Bottles without Closure: Bulk Pack

Safely store and handle test kits and many small volume applications with these PPCO diagnostic bottles, for use in sample storage and shipping small volume reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Neck Finish
362006-91253.4 mL13-415 mm
362006-90258 mL20-415 mm
362006-905015 mL20-415 mm
Catalog number 362006-9125
Price (USD)
849.47
Online exclusive
999.38
Save 149.91
Case of 2000
Capacity (Metric):
3.4 mL
Neck Finish:
13-415 mm
Price (USD)
849.47
Online exclusive
999.38
Save 149.91
Case of 2000

Safely store and handle test kits and many small volume applications with Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles, for use in sample storage and the shipping of small volume, valuable reagents. These polypropylene copolymer bottles are durable and feature uniform walls resistant to splitting or puncturing. They are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).

Features:
  • Nonsterile
  • Natural translucent PPCO provides excellent chemical resistance
  • Autoclavable
  • Leakproof† when assembled with closure (must be ordered separately)

Ordering Information:

Bottles are bulk packed without closures. Packed 2000 per case. Order colored closures separately (Cat. No. 362150).

.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
AutoclavableAutoclavable
Capacity0.125 oz., 3.4 mL
Capacity (English)0.125 oz.
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Diameter (Metric)16 mm
Diameter (Metric) Inner Neck8 mm
Height (English)1.61 in.
Height (Metric)41 mm
Neck Finish13-415 mm
No. per Case2000
No. per Pack2000
Product TypeSample Container
Through the Closure DispensingNo
Width (English)0.62 in.
Capacity (Metric)3.4 mL
Closure IncludedYes
ColorTranslucent
MaterialPPCO
MouthNarrow
SterilityNon-sterile
Unit SizeCase of 2000

Frequently asked questions (FAQs)

Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.