Streptavidin - HRP
Invitrogen™

Streptavidin - HRP

Streptavidin - HRP
Have Questions?
Catalog NumberQuantity
4343232.5 mg
Catalog number 434323
Price (EUR)
456,00
Each
Quantity:
2.5 mg
Request bulk or custom format
Price (EUR)
456,00
Each

Reactivity: HRP-SA reacts with biotin.

Applications: Immunohistochemistry, Western blotting, and ELISA

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Concentration1.25 mg/mL
Label or DyeHRP (Horseradish Peroxidase)
Product TypeStreptavidin Conjugate (fluorescent)
Quantity2.5 mg
Shipping ConditionWet Ice
ConjugateHRP
FormLiquid
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).

Citations & References (11)

Citations & References
Abstract
Proteoglycan abnormalities in olfactory epithelium tissue from subjects diagnosed with schizophrenia.
Authors:Pantazopoulos H, Boyer-Boiteau A, Holbrook EH, Jang W, Hahn CG, Arnold SE, Berretta S
Journal:
PubMed ID:24035561
'Emerging evidence points to proteoglycan abnormalities in the pathophysiology of schizophrenia (SZ). In particular, markedly abnormal expression of chondroitin sulfate proteoglycans (CSPGs), key components of the extracellular matrix, was observed in the medial temporal lobe. CSPG functions, including regulation of neuronal differentiation and migration, are highly relevant to the pathophysiology ... More
Histopathological changes in the upper respiratory tract of F344 rats following infection with a rat-adapted influenza virus.
Authors:Ibanes JD, Morgan KT, Burleson GR
Journal:Vet Pathol
PubMed ID:8817839
'The present study determined the morphogenesis of upper respiratory tract disease in rats following infection with a rat-adapted influenza virus. Sixty-eight 60-day-old, male F344 rats were infected by intranasal inoculation and necropsied at days 1, 2, 4, 7, 14, and 28 post-inoculation (PI). Responses to infection were studied by routine ... More
A locus control region adjacent to the human red and green visual pigment genes.
Authors:Wang Y, Macke JP, Merbs SL, Zack DJ, Klaunberg B, Bennett J, Gearhart J, Nathans J
Journal:Neuron
PubMed ID:1524826
'Deletion of sequences 5'' of the human red and green pigment gene array results in blue cone monochromacy, a disorder in which both red and green cone function are absent. To test whether these sequences are required for transcription of the adjacent visual pigment genes in cone photoreceptors, we produced ... More
Fas ligand is positioned in mouse uterus and placenta to prevent trafficking of activated leukocytes between the mother and the conceptus.
Authors:Hunt JS, Vassmer D, Ferguson TA, Miller L
Journal:J Immunol
PubMed ID:9126971
'Despite intimate juxtaposition of maternal and fetal tissues during mammalian pregnancy, reciprocal migration of cells is limited. To evaluate the postulate that cell traffic is restricted by expression of Fas ligand (FasL) in the uterus and placenta, FasL mRNA was identified by using reverse transcription-PCR, and FasL protein was identified ... More
The neutrophil selectin LECAM-1 presents carbohydrate ligands to the vascular selectins ELAM-1 and GMP-140.
Authors:Picker LJ, Warnock RA, Burns AR, Doerschuk CM, Berg EL, Butcher EC
Journal:Cell
PubMed ID:1716182
'LECAM-1 (leukocyte-endothelial cell adhesion molecule 1), the lymphocyte lectin ("selectin") homing receptor for peripheral lymph nodes (PLNs), participates in the earliest interactions of polymorphonuclear neutrophilic leukocytes (PMNs) with inflamed venules. Here, we present evidence that LECAM-1 mediates this function through a novel mechanism--presentation of oligosaccharide ligands to the inducible vascular ... More