MagMAX™ Pathogen RNA/DNA Kit - FAQs
View additional product information for MagMAX™ Pathogen RNA/DNA Kit - FAQs (4462359)
8 product FAQs found
What type of samples can I use with the MagMAX Pathogen RNA/DNA kit?
The MagMAX Pathogen RNA/DNA Kit is designed for rapid purification of nucleic acid (RNA and DNA) from the following sample types:
- Low-cell content samples including serum, plasma, swabs (nasal, tracheal, and cloacal), ear notches
- Whole blood
- Semen
- Oral fluid
- Feces
- Biomed Diagnostics InPouch TF (Tritrichomonas foetus) culture
Do you have a protocol for manual purification using a magnetic stand with the MagMAX Pathogen RNA/DNA Kit?
The manual isolation protocol for the MagMax Pathogen RNA/DNA Kit can be found in the MagMAX Pathogen RNA/DNA Kit Protocol , page 49.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.
Which beads should I use with the MagMax Pathogen RNA/DNA Kit (Cat. No. 4462359)?
We recommend using DNA Binding Beads for MagMAX-96 DNA Multi-Sample Kit (Cat. No. 4489112) with the MagMax Pathogen RNA/DNA Kit.
Can I edit the manufactured KingFisher Flex protocols?
No, manufactured protocols are locked for editing, but they can be renamed using the ‘save as' function and edited as a new version.
Can I use magnetic beads from other vendors with the KingFisher Flex instrument?
For nucleic acid extraction, we recommend using our MagMAX kits and MagJET kits. Other magnetic beads should work well as long as they are 0.5 to 10 µm in size. However, you will have to write your own KingFisher protocols using the Thermo Scientific BindIt software and validate/optimize the protocol on your own.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.
I am working with liquid samples with high volume and want to extract nucleic acid from them using the KingFisher Flex instrument. Can I use more volume than the protocol advises?
Yes, this can be done by sample splitting. Set the protocol so that the magnet will collect samples from multiple wells before continuing to the next step. Be sure not to overload.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.
Can I scale up my reaction volume with the KingFisher Flex instrument?
Yes, reaction volumes can be scaled up, and all other volumes (i.e. wash volumes) will have to be increased by the same fold difference. It is generally not recommended to scale up reaction volumes more than 2-fold unless the plate's volume capacity will allow it.
Are all of the Applied Biosystems MagMAX kits validated for the KingFisher Flex Magnetic Particle Processor?
Yes, all MagMAX kits can be used with the KingFisher Flex Magnetic Particle Processor.