Pierce™ Protease Inhibitor Tablets, EDTA-free
Thermo Scientific™
Pierce™ Protease Inhibitor Tablets, EDTA-free
Thermo Scientific Pierce Protease Inhibitor Tablets, EDTA-free, contain broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation duringRead more
Have Questions?
Catalog NumberQuantity
A3296520 Tablets
Catalog number A32965
Price (USD)
350.65
Online exclusive
403.00
Save 52.35
Each
Quantity:
20 Tablets
Price (USD)
350.65
Online exclusive
403.00
Save 52.35
Each
Thermo Scientific Pierce Protease Inhibitor Tablets, EDTA-free, contain broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation during cell lysis and protein extraction experiments.
Features of EDTA-free Protease Inhibitor Tablets:
• Improved formulation—tablets dissolve quickly to form a clear solution
• Economical—more cost effective than other commercially available formulations for equivalent volumes
• Convenient—ready-to-use, fully disclosed, broad-spectrum formulations
• Compatible—use directly with Pierce BCA assays
• EDTA-free—formulated without EDTA, a metalloproteinase inhibitor (only for EDTA-free formulations)
Pierce Protease Inhibitor Tablets protect proteins from proteolytic degradation through inhibition of the serine proteases, cysteine proteases, aspartic acid proteases, and aminopeptidases that are typically present in cell lysate samples. The tablets contain AEBSF, aprotinin, bestatin, E-64, leupeptin, and pepstatin A. Stable for storage at 4°C for up to 12 months, each tablet is sufficient for 50 mL of solution, and is suitable for reagent preparation prior to protein extraction from tissue and cultured cells.
Proteases make up a large category of enzymes, including endopeptidases and exopeptidases, which catalyze the hydrolytic breakdown of proteins into peptides or amino acids. Proteases reside in cellular compartments, but upon cell lysis the proteases are released, resulting in the degradation of valuable proteins. For this reason, it is common practice to add multiple compounds known to inactivate or inhibit proteases to cell lysis buffers or cell extracts.
To use Pierce Protease Inhibitor Tablets, simply dissolve 1 tablet in 50 mL of buffer or lysate. The protease inhibitor formulation is compatible with most detergent-based cell lysis reagents, including Pierce Cell Lysis solutions. Pierce Protease Inhibitor Tablets do not contain phosphatase inhibitors. If necessary, phosphatase inhibitors, such as Pierce Phosphatase Inhibitor Mini Tablets, can be added along with the protease inhibitor tablets.
Related products
Pierce Protease Inhibitor Tablets
Pierce Protease Inhibitor Mini Tablets, EDTA-free
Pierce Phosphatase Inhibitor Mini Tablets
Pierce Protease and Phosphatase Inhibitor Mini Tablets
For Research Use Only. Not for use in diagnostic procedures.
Specifications
InhibitorsProtease Inhibitor
Quantity20 Tablets
Shipping ConditionRoom Temperature
Sufficient For50 mL Solution
FormSolid
Product LinePierce
Product TypeProtease Inhibitor Cocktail
Unit SizeEach
Contents & Storage
1 Vial, Store at 2°C to 8°C
Frequently asked questions (FAQs)
Can I use just part of the Pierce Protease Inhibitor XL Capsules, EDTA-Free and save part of the capsule for later use?
We do not recommend “splitting” the capsule into smaller portions. Researchers using less than 500 mL should choose a different format (liquid or tablet) of protease inhibitors.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What formats of protease inhibitors do you offer?
We offer standalone protease inhibitors (AEBSF, Aprotinin, PMSF, EDTA, etc.), as well as cocktails or combinations of protease inhibitors in liquid, tablet, and larger capsule formats. These cocktails of protease inhibitors can also be purchased in combination with a cocktail of phosphatase inhibitors, in both liquid and tablet form, for even greater convenience.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Do you offer any of the components of the Pierce protease, phosphatase and (protease and phosphatase) inhibitor tablets and mini tablets individually?
Yes, the following components of the Pierce protease, phosphatase and (protease and phosphatase) inhibitor tablets and mini tablets are offered separately:
- AEBSF (Cat. No. 78431)
- Aprotinin (Cat. No. 78432)
- Bestatin (Cat. No. 8433)
- E-64 (Cat. No. 78434)
- Leupeptin (Cat. No. 78435)
- Pepstatin A (Cat. No. 78436)
- EDTA (Cat. No. 17892).
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What are the concentrations of the various components of the protease and phosphatase inhibitor tablets and mini tablets?
The final concentration of each component in the protease and phosphatase inhibitor tablets and mini tablets is proprietary. We release only the identities of the inhibitors.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Can I store the reconstituted solution of protease, phosphatase and (protease and phosphatase) inhibitor tablet or mini tablet and if so, for how long?
The Pierce protease, phosphatase, as well as (protease and phosphatase) inhibitor tablets and mini tablets are stable for at least one year when stored at 4 degrees C, but we only recommend preparing fresh solutions just before needed. If you are interested in liquid protease or phosphatase inhibitors, please use our inhibitor cocktails (https://www.thermofisher.com/search/results?query=protease%20inhibitor%20cocktails&focusarea=Search%20All).
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Citations & References (3)
Citations & References
Abstract
An Introductory Guide to Protease Sensitive Linker Design Using Matrix Metalloproteinase 13 as an Example.
Authors:Hamm P,Meinel L,Driessen MD
Journal:ACS biomaterials science & engineering
PubMed ID:38813796
Proteases play a crucial role, not only in physiological, but also in pathological processes, such as cancer, inflammation, arthritis, Alzheimer's, and infections, to name but a few. Their ability to cleave peptides can be harnessed for a broad range of biotechnological purposes. To do this efficiently, it is essential to ... More
Systematic comparison of CRISPR-based transcriptional activators uncovers gene-regulatory features of enhancer-promoter interactions.
Authors:Wang K,Escobar M,Li J,Mahata B,Goell J,Shah S,Cluck M,Hilton IB
Journal:Nucleic acids research
PubMed ID:35849129
Nuclease-inactivated CRISPR/Cas-based (dCas-based) systems have emerged as powerful technologies to synthetically reshape the human epigenome and gene expression. Despite the increasing adoption of these platforms, their relative potencies and mechanistic differences are incompletely characterized, particularly at human enhancer-promoter pairs. Here, we systematically compared the most widely adopted dCas9-based transcriptional activators, ... More
Not just fat: investigating the proteome of cetacean blubber tissue.
Authors:Kershaw JL,Botting CH,Brownlow A,Hall AJ
Journal:Conservation physiology
PubMed ID:29479430
Mammalian adipose tissue is increasingly being recognized as an endocrine organ involved in the regulation of a number of metabolic processes and pathways. It responds to signals from different hormone systems and the central nervous system, and expresses a variety of protein factors with important paracrine and endocrine functions. This ... More