CTS™ OpTmizer™ Pro SFM, bottle format - FAQs
CTS™ OpTmizer™ Pro SFM, bottle format - FAQs
View additional product information for CTS™ OpTmizer™ Pro SFM, bottle format - FAQs (A4966101)
2 product FAQs found
What is CTS?
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
What is the primary advantage of CTS OpTmizer Pro SFM over CTS OpTmizer T-Cell Expansion SFM?
CTS OpTmizer Pro SFM has a proprietary formulation that has been demonstrated to increase the growth and expansion of central memory T-cell phenotypes that may be more desirable in allogeneic T cell therapy workflows.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.