Search
Search
Bethyl Laboratories
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
SMARCA2 encodes a protein that is a member of the SWI/SNF family of proteins and is highly similar to the brahma protein of Drosophila. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI, which is required for transcriptional activation of genes normally repressed by chromatin. Two transcript variants encoding different isoforms have been found for this gene, which contains a trinucleotide repeat (CAG) length polymorphism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent helicase SMARCA2; BAF190B; brahma homolog; BRG1-associated factor 190B; FLJ36757; global transcription activator homologous sequence; MGC74511; N-terminal region of the protein; probable global transcription activator SNF2L2; protein brahma homolog; putative global transcription activator SNF2L2; SNF2-alpha; SNF2/SWI2-like protein 2; subunit of the SWI/SNF chromatin remodeling complex; sucrose nonfermenting 2-like protein 2; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin a2; unnamed protein product
Gene Aliases: 2610209L14Rik; BAF190; BIS; brahma; BRM; hBRM; hSNF2a; NCBRS; SAMRCA2; SNF2; SNF2alpha; SNF2L2; SNF2LA; Sth1p; SWI2
UniProt ID: (Human) P51531, (Mouse) Q6DIC0
Entrez Gene ID: (Human) 6595, (Mouse) 67155
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support