Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The Retinol Binding Protein Competitive ELISA quantitates retinol binding protein (RBP) in serum, plasma, urine, saliva, dried fecal extracts or cell culture medium.|Principle of the method: The Retinol Binding Protein Competitive ELISA research-use-only kit is designed to quantitatively measure RBP independent of species. An RBP standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with anti-sheep IgG antibodies. Anti-sheep RBP antibodies and RBP-peroxidase conjugate is added to the wells. The RBP-peroxidase conjugate and any RBP in the sample will compete to bind to the anti-sheep RBP antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound RBP-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of RBP present in the sample.|Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
This gene encodes the carrier protein involved in the transport of retinol (vitamin A alcohol) from the liver storage site to peripheral tissue. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : CRABP-I, CRBP, CRBP1, CRBPI, hCRBP1, RBPC
Gene ID : (Human) 5947
Gene symbol : RBP1
Protein Aliases : C CRABP-I, RBP, retinol, retinol binding, Retinol Binding Protein, cellular retinol binding protein 1, cellular retinol-binding protein I, RBP1, retinol-binding protein, retinol-binding protein 1, cellular, unnamed protein product
UniProt ID (Human) P09455
If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support