Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGGSGSRLSK ELLAEYQDLT FLTKQEILLA HRRFCELLPQ EQRSVESSLR AQVPFEQILS LPELKANPFK ERICRVFSTS PAKDSLSFED FLDLLSVFSD TATPDIKSHY AFRIFDFDDD GTLNREDLSR LVNCLTGEGE DTRLSASEMK QLIDNILEES DIDRDGTINL SEFQHVISRS PDFASSFKIV L
The protein encoded by this gene is a member of the calcium-binding protein family. The specific function of this protein has not yet been determined; however this protein is known to interact with DNA-dependent protein kinase and may play a role in kinase-phosphatase regulation of DNA end joining. This protein also interacts with integrin alphabeta, which may implicate this protein as a regulatory molecule for alphabeta.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium and integrin binding 1 (calmyrin); Calcium and integrin-binding protein 1; Calcium- and integrin-binding protein; calcium-binding protein family member; CIB1; Calmyrin; CIB; CIB1; CIBP; DNA-dependent protein kinase interacting protein; DNA-PK interaction protein; DNA-PKcs-interacting protein; Kinase-interacting protein; KIP; SIP2-28; SNK-interacting protein 2-28; testicular secretory protein Li 9
Gene Aliases: CIB; CIB1; CIBP; EV3; KIP; KIP1; PRKDCIP; SIP2-28
UniProt ID: (Human) Q99828
Entrez Gene ID: (Human) 10519
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support