How can I reconstitute Protease Inhibitor Cocktail II?
Protease Inhibitor Cocktail II is recommended for use with bacterial cell extracts. You should first reconstitute each vial in 1 mL DMSO. Then add 4 mL DI water to obtain a 5 mL stock solution. The solution may appear turbid. The 5 mL stock is enough to inhibit protease activity extracted from a 20 g bacterial cell pellet.
How do I prepare the Protease Inhibitor Cocktail II?
We recommend that you reconstitute each vial with 1 mL DMSO first, and then add 4 mL water to obtain a 5 mL stock solution. The solution may appear turbid after reconstitution. 5 mL solution is recommended for the inhibition of proteases extracted from 20 g of E. coli. A 5 mL solution contains the following: 20 mM AEBSF.HCl, 1.7 mM bestatin, 200 µM E-64, 85 mM EDTA, and 2 mM pepstatin A.