Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
It is not recommended to aliquot this antibody.
This antibody is not cross-reactive with other related proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-AMP-activated protein kinase 40 kDa subunit; 5'-AMP-activated protein kinase beta-1 subunit; 5'-AMP-activated protein kinase subunit beta-1; 5'-AMP-activated protein kinase subunit beta-1-like protein; 5'-AMP-activated protein kinase subunit beta-2; 5'-AMP-activated protein kinase, beta subunit; 5'-AMP-activated protein kinase, beta-2 subunit; 5-AMP-activated protein kinase beta subunit; AMP-activated protein kinase beta 2 non-catalytic subunit; AMP-activated protein kinase beta subunit; AMP-activated protein kinase beta-2 regulatory subunit; AMPK beta -1 chain; AMPK beta 1; AMPK beta 2; AMPK beta-1 chain; AMPK beta-2 chain; AMPK subunit beta-1; AMPK subunit beta-2; AMPKb; protein kinase, AMP-activated, beta 1 non-catalytic subunit; protein kinase, AMP-activated, beta 2 non-catalytic subunit; protein kinase, AMP-activated, noncatalytic, beta-1; unnamed protein product
Gene Aliases: 1300015D22Rik; 5730553K21Rik; AMPK; AU021155; AW049591; BB124140; E430008F22; HAMPKb; I79_012592; PRKAB1; PRKAB2
UniProt ID: (Human) Q9Y478, (Human) O43741, (Mouse) Q9R078, (Rat) P80386, (Rat) Q9QZH4, (Mouse) Q6PAM0
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support