Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Positive control: Mouse eyeball tissue lysate, rat eyeball tissue lysate.
Subcellular location: Cytoplasm, cytosol, Nucleus, Cell projection, cilium, photoreceptor outer segment, Photoreceptor inner segment.
Recommended dilution for IF-tissue samples: 1:200-1:2,000
PDC encodes a phosphoprotein, which is located in the outer and inner segments of the rod cells in the retina. This protein may participate in the regulation of visual phototransduction or in the integration of photoreceptor metabolism. It modulates the phototransduction cascade by interacting with the beta and gamma subunits of the retinal G-protein transducin. PDC is a potential candidate gene for retinitis pigmentosa and Usher syndrome type II. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 33 kDa phototransducing protein; 33-kDa phototransducing protein; G beta gamma binding protein; PHD; phosducin-like orphan protein; Phototransducing protein, 33 kDa (phosducin); rod photoreceptor 1; RPR-1
Gene Aliases: 33DPTP; MEKA; PHD; PhLOP; PhLP; Rpr-1; Rpr1
UniProt ID: (Human) P20941, (Mouse) Q9QW08, (Rat) P20942
Entrez Gene ID: (Human) 5132, (Mouse) 20028, (Rat) 25343
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support