Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. This structure consists of approximately 146 bp of DNA wrapped around a nucleosome, an octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is found in a histone cluster on chromosome 1. This gene is one of four histone genes in the cluster that are duplicated; this record represents the telomeric copy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: di-methyl H3; di-methyl histone H3; Di-methyl-Histone H3; dimethyl H3; dimethyl histone H3; Dimethyl-Histone H3; H3 histone family, member L; H3-clustered histone 13; H3-clustered histone 14; H3-clustered histone 15; H3-clustered histone 2; H3-clustered histone 3; H3-clustered histone 4; H3-clustered histone 6; H3-clustered histone 7; H3Arg8me2s; histone 1, H3b; histone 2, H3a; histone 2, H3c2; histone 2, H3ca2; histone cluster 1 H3 family member b; histone cluster 1, H3b; histone cluster 2 H3 family member a; histone cluster 2 H3C family member 2; histone cluster 2, H3a; histone cluster 2, H3c2; histone cluster 2, H3c2, pseudogene; Histone H3; histone H3/l; histone H3/o
Gene Aliases: H3-53; H3-614; H3.2-614; H3/l; H3/n; H3/o; H3c13; H3c14; H3c15; H3c2; H3c3; H3c4; H3c6; H3c7; H3FL; HIST1H3B; HIST2H3A; Hist2h3c2; Hist2h3c2-ps; Hist2h3ca1
UniProt ID: (Human) Q71DI3, (Human) P68431, (Mouse) P84228
Entrez Gene ID: (Human) 333932, (Human) 8358, (Mouse) 97114, (Rat) 310678, (Mouse) 319150
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support