The green fluorescent fluorescein ovalbumin conjugate is a protein with a relatively low molecular weight (∼45,000 daltons) labeled with fluorescein.자세히 알아보기
Have Questions?
카탈로그 번호
수량
O23020
5 mg
카탈로그 번호 O23020
제품 가격(KRW)
231,000
온라인 행사
Ends: 31-Dec-2026
264,000
할인액 33,000 (13%)
Each
수량:
5 mg
제품 가격(KRW)
231,000
온라인 행사
Ends: 31-Dec-2026
264,000
할인액 33,000 (13%)
Each
The green fluorescent fluorescein ovalbumin conjugate is a protein with a relatively low molecular weight (∼45,000 daltons) labeled with fluorescein.
For Research Use Only. Not for use in diagnostic procedures.
사양
라벨 유형Classic Dyes
단백질 아형Albumin Proteins
수량5 mg
배송 조건Room Temperature
콘주게이트Fluorescein
형태Lyophilized
Unit SizeEach
구성 및 보관
Store in freezer (-5 to -30°C) and protect from light.
인용 및 참조 문헌 (46)
인용 및 참조 문헌
Abstract
The microvasculature of the hamster cheek pouch as a model for studying acute immune-complex-induced inflammatory reactions (with 1 color plate).
Authors:Björk J, Smedegård G
Journal:Int Arch Allergy Appl Immunol
PubMed ID:6232222
'The acute microvascular reactions induced by immune complexes were studied. Hamster cheek pouch preparations of ovalbumin (OA)-immunized animals were topically exposed to different doses of OA (0.1, 1, 10 or 100 micrograms/ml). Fluorescein-isothiocyanate-labelled dextran (FITC-dextran, molecular weight 150,000) was injected intravenously as a macromolecular tracer. The microvascular alterations observed upon ... More
Low molecular weight antigen arrays delete high affinity memory B cells without affecting specific T-cell help.
Authors:Reim JW, Symer DE, Watson DC, Dintzis RZ, Dintzis HM
Journal:Mol Immunol
PubMed ID:9171897
'An ongoing, T-cell dependent, secondary antibody response to an epitope can be suppressed in vivo by low molecular weight, soluble polymers, bearing multiple copies of the same epitope. This study illustrates that such suppressive T-cell independent antigen arrays target the epitope-specific, high affinity, memory B cells for long-term functional elimination. ... More
Permeability of the primary decidual zone in the rat uterus: studies using fluorescein-labeled proteins and dextrans.
Authors:Parr MB, Parr EL
Journal:Biol Reprod
PubMed ID:2420379
'The primary decidual zone (PDZ) is a transitory avascular region of transformed fibroblasts surrounding the luminal epithelium at the implantation site. Since this zone may restrict the passage of immunoglobulins, cells, nutrients, and other substances from maternal blood to the epithelium and embryo from Days 6 to 8 of pregnancy, ... More
Hepatic sinusoidal endothelium heterogeneity with respect to mannose receptor activity is interleukin-1 dependent.
Authors:Asumendi A, Alvarez A, Martinez I, Smedsrød B, Vidal-Vanaclocha F
Journal:Hepatology
PubMed ID:8675173
'Using fluorescein isothiocyanate-conjugated ovalbumin (OVA-FITC), 125I-mannan, or 125I-invertase as specific ligands for the mannose receptor, we have quantified its activity in mouse and rat hepatic sinusoidal endothelium (HSE), under both basal conditions and after lipopolysaccharide (LPS) or human recombinant interleukin-1beta (IL-1beta) stimulations. Mouse treatment for 4 hours with 5 microg/kg ... More
Preparation of fluorescent derivatives of lipases and their use in fluorescence energy transfer studies in hydrocarbon/water interfaces.
Authors:Roberts GA, Tombs MP
Journal:Biochim Biophys Acta
PubMed ID:3113482
'Fluorescein isothiocyanate reacted with a chromobacter and pseudomonad lipase to yield mono-substituted, fully active, enzymes. With the carbocyanine dye 1,1''-dioctadecyl-3,3,3'',3''-tetramethylindocarbocyanine perchlorate (DiI) in the non-aqueous phase, fluorescence energy transfer was used to follow the lipase and similarly labelled model proteins in and out of the interface in heptane, and heptane/di-O-palmitoyl-rac-glycerol ... More