Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: LFYLTNSEYR SEQSVKLRRQ VIQVVLNGME SYQEVTVQRN CCLTLCNFSI PEELEFQYRR VNELLLSILN PTRQDESIQR IAVHLCNALV C
Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.
ZER1 probably acts as target recruitment subunit in the E3 ubiquitin ligase complex ZER1-CUL2-Elongin BC.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homolog of Zyg-11; RP11-545E17.4; unnamed protein product; zer-1 homolog; ZYG homolog; zyg-11 homolog B-like protein; zyg11b-like protein
Gene Aliases: C9orf60; ZYG; ZYG11BL
UniProt ID: (Human) Q7Z7L7
Entrez Gene ID: (Human) 10444
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support