Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: LDRLYAQRPA GFGCQESRHS YPPALGSPGA LAGAGVGAAG PLERRGAQPG RHSVTGYGDC AVGARYQDEL TALLRLTVGT GGREAGARGE PSGIEPSGLE EPPGPFVPEA ARARMREPEA REDYFGTCIK CNKGIYGQSN
Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.
AJUBA (Ajuba LIM Protein) is a Protein Coding gene. Diseases associated with AJUBA include Wilson-Turner X-Linked Mental Retardation Syndrome and Syndromic X-Linked Intellectual Disability. Among its related pathways are Hippo signaling pathway and Adhesion. Gene Ontology (GO) annotations related to this gene include chromatin binding and actin filament binding. An important paralog of this gene is LIMD1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: jub, ajuba homolog; LIM domain containing protein; unnamed protein product
Gene Aliases: JUB
UniProt ID: (Human) Q96IF1
Entrez Gene ID: (Human) 84962
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support