Search
Search
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: FIYTREGRPS GEAFVELESE EEVKLALKKD RETMGHRYVE VFKSNSVEMD WVLKHTGPNS PDTANDGFVR LRGLPFGCSK EEIVQFFSGL EIVPNGMTLP VDFQGRSTGE AFVQFAS
Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that binds to RNAs. It is very similar to the family member HNRPH1. This gene is thought to be involved in Fabray disease and X-linked agammaglobulinemia phenotype. Alternative splicing results in multiple transcript variants encoding the same protein. [provided by RefSeq].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory sperm binding protein; FTP-3; heterogeneous nuclear ribonucleoprotein H-prime; heterogeneous nuclear ribonucleoprotein H2 (H'); hnRNP H'; hnRNP H2; unnamed protein product
Gene Aliases: FTP3; hnRNPH'; HNRPH'; HNRPH2; MRXSB; NRPH2
UniProt ID: (Human) P55795
Entrez Gene ID: (Human) 3188
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support