Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: DAEGKLWVAC YNGGRVIRLD PVTGKRLQTV KLPVDKTTSC CFGGKNYSEM YVTCARDGMD PEGLLRQPEA GGIFKITGLG VKGIAPYSY
Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
The protein encoded by this gene is a highly conserved, calcium-binding protein, that is preferentially expressed in the liver and kidney. It may have an important role in calcium homeostasis. Studies in rat indicate that this protein may also play a role in aging, as it shows age-associated down-regulation. This gene is part of a gene cluster on chromosome Xp11.3-Xp11.23. Alternative splicing results in two transcript variants having different 5' UTRs, but encoding the same protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 41; gluconolactonase; regucalcin (senescence marker protein-30); senescence marker protein 30; SMP-30; unnamed protein product
Gene Aliases: GNL; HEL-S-41; RC; Reguc; SMP-30; SMP30
UniProt ID: (Human) Q15493, (Rat) Q03336, (Mouse) Q64374
Entrez Gene ID: (Human) 9104, (Rat) 25106, (Mouse) 19733
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support