Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: EEHIPHQQIQ AQLVAGQSLA GGQQIQIQTV GALSPPPSQQ GSPREGERRV GTASVLQPVK KRKVDMPITV SYAISGQPVA TVLAIPQGQ
Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Glutamine-rich protein 1 contains 1 CARD domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20259; glutamine-rich protein 1; MGC131838; unnamed protein product
Gene Aliases: 2610028H07Rik; AB-DIP; b2b2404Clo; RGD1311095; VERBRAS; VERBRAS1
UniProt ID: (Human) Q2TAL8, (Mouse) Q3UA37
Entrez Gene ID: (Human) 54870, (Mouse) 69232, (Rat) 301004
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support