Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: IMLGKQLEGI WHTSIVVHKD EFFFGSGGIS SCPPGGTLLG PPDSVVDVGS TEVTEE
Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Protease which deconjugates SUMO1, SUMO2 and SUMO3 from some substrate proteins. Has isopeptidase but not SUMO-processing activity. Desumoylates ZBTB46. Collaborates with UBQLN4 in the export of ubiquitinated proteins from the nucleus to the cytoplasm (PubMed:29666234). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dJ347H13.4; family with sequence similarity 152, member B; palmitoyl protein thioesterase DESI1; polyubiquitinated substrate transporter; PPPDE peptidase domain containing 2; PPPDE peptidase domain-containing protein 2; S-depalmitoylase DESI1
Gene Aliases: D15Wsu75e; DeSI-1; DJ347H13.4; FAM152B; POST; PPPDE2
UniProt ID: (Human) Q6ICB0
Entrez Gene ID: (Human) 27351
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support