Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: FYKVPRSQTK LTIMLEKLGM DYDGRPHCGL DDSKNIARIA VRMLQDGCEL RINEKMHAGQ LMSVSSS
Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 96%.
RNA exonuclease that binds to the 3'-end of histone mRNAs and degrades them, suggesting that it plays an essential role in histone mRNA decay after replication. A 2' and 3'-hydroxyl groups at the last nucleotide of the histone 3'-end is required for efficient degradation of RNA substrates. Also able to degrade the 3'-overhangs of short interfering RNAs (siRNAs) in vitro, suggesting a possible role as regulator of RNA interference (RNAi). Requires for binding the 5'-ACCCA-3' sequence present in stem-loop structure. Able to bind other mRNAs. Required for 5.8S rRNA 3'-end processing. Also binds to 5.8s ribosomal RNA. Binds with high affinity to the stem-loop structure of replication-dependent histone pre-mRNAs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3' exoribonuclease; 3'-5' exonuclease ERI1; 3'hExo; enhanced RNAi three prime mRNA exonuclease homolog 1; histone mRNA 3' end-specific exonuclease; histone mRNA 3'-end-specific exoribonuclease; histone mRNA 3'-exonuclease 1; protei; three prime histone mRNA exonuclease 1; three prime mRNA exonuclease 1; unnamed protein product
Gene Aliases: 3'HEXO; HEXO; HXAL; SEMDGC; SEMDGS; THEX1
UniProt ID: (Human) Q8IV48
Entrez Gene ID: (Human) 90459
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support