Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 6
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: VLRHVNGQDQI VPGLYACGEA ACASVHGANR LGANSLLDLV VFGRACALSI EESCRPGDKV PPIKPNAGEE S
Highest antigen sequence identity to the following orthologs - mouse 94%, rat 96%.
This gene encodes a major catalytic subunit of succinate-ubiquinone oxidoreductase, a complex of the mitochondrial respiratory chain. The complex is composed of four nuclear-encoded subunits and is localized in the mitochondrial inner membrane. Mutations in this gene have been associated with a form of mitochondrial respiratory chain deficiency known as Leigh Syndrome. A pseudogene has been identified on chromosome 3q29.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: flavoprotein; flavoprotein subunit of complex II; malate dehydrogenase [quinone] flavoprotein subunit; succinate dehydrogenase complex, subunit A, flavoprotein (Fp); unnamed protein product
Gene Aliases: CMD1GG; FP; MC2DN1; NDAXOA; PGL5; PPGL5; SDH1; SDH2; SDHF
UniProt ID: (Human) P31040
Entrez Gene ID: (Human) 6389
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support