Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: HLKKPSQLSS FSWDNCDEGK DPAVIRSLTL EPDPIIVPGN VTLSVMGSTS VPLSSPLKVD LVLEKEVAGL WIKIPCTDYI GSCTFEHFCD VLDMLIPTGE PCPEPLRTYG LPCHCPFKEG TYSLPKSEFV VPDLELPSWL TTGNYRIESV LSSSGKRLGC IKIAASLKGI
This gene encodes a small glycolipid transport protein which acts as a substrate specific co-factor for the lysosomal enzyme beta-hexosaminidase A. Beta-hexosaminidase A, together with GM2 ganglioside activator, catalyzes the degradation of the ganglioside GM2, and other molecules containing terminal N-acetyl hexosamines. Mutations in this gene result in GM2-gangliosidosis type AB or the AB variant of Tay-Sachs disease. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cerebroside sulfate activator protein; GM2 ganglioside activator; GM2 ganglioside activator protein; GM2-activator protein; Gm2a; shingolipid activator protein 3; sphingolipid activator protein 3; unnamed protein product
Gene Aliases: GM2-AP; GM2AP; SAP-3
UniProt ID: (Human) P17900, (Mouse) Q60648
Entrez Gene ID: (Human) 2760, (Mouse) 14667, (Rat) 282838
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support