Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch and glycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas. It is an amylase isoenzyme produced by the salivary gland.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; A-1 alpha-amylase; a-amylase; alpha-amylase; amylase 2, pancreatic; amylase 2-1, pancreatic; amylase 2-2, pancreatic; amylase 2a1, pancreatic; amylase 2a3, pancreatic; amylase 2b, pancreatic; amylase alpha 2A (pancreatic); amylase alpha 2B (pancreatic); B-1 alpha-amylase; carcinoid alpha-amylase; glycogenase; PA; Pancreatic alpha-amylase; pancreatic alpha-amylase-like; pancreatic amylase; pancreatic amylase alpha 2A; unnamed protein product
Gene Aliases: Amy-X; AMY2; Amy2-1; Amy2-2; Amy2a; AMY3; HXA; mAmy2-1; mAmy2-4; mAmy2-6; PA
UniProt ID: (Human) P04746, (Human) P19961, (Rat) P00689, (Mouse) P00688
Entrez Gene ID: (Human) 279, (Human) 280, (Mouse) 100043207, (Rat) 497039, (Mouse) 100043686, (Mouse) 545562
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support