Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Positive Control - IHC: human mammary cancer tissue, mouse testis tissue.
DRB1 is RNA-binding protein with binding specificity for poly(C) and may play an important role in neural development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: developmentally regulated RNA binding protein 1; developmentally regulated RNA-binding protein 1; dopamine receptor binding 1; putative RNA binding protein RB-1; RB-1; unnamed protein product
Gene Aliases: DRB1; Drbp1; G430095G15Rik; RB-1
UniProt ID: (Human) Q8IUH3, (Mouse) Q8BHN5
Entrez Gene ID: (Human) 129831, (Mouse) 241490
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support