Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
ABCA9 (ATP-binding cassette, sub-family A (ABC1), member 9) is a 1,624 amino acid protein belonging to the ABC transporter superfamily and the ABCA family. The ABC1 subfamily is the only major ABC subfamily exclusive to multicellular eukaryotes. Ubiquitously expressed, with high expression in heart, brain and fetal tissues, ABCA9 is a multi-pass membrane protein that contains two ABC transporter domains and exists as five alternatively spliced isoforms. ABCA9 exhibits membrane subcellular localization and may play a role in monocyte differentiation and lipid homeostasis. ABCA9 is up-regulated during monocyte differentiation into macrophages and down-regulated by cholesterol loading of macrophages. Spanning 85 kb, ABCA9 contains 39 exons, with a non-coding first exon.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABC-A9; ABCA9; ATP-binding cassette A9; ATP-binding cassette, sub-family A (ABC1), member 9; DKFZp686F2450; MGC75415; unnamed protein product
Gene Aliases: EST640918
UniProt ID: (Human) Q8IUA7
Entrez Gene ID: (Human) 10350
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support