Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Recommended positive controls: A431, HeLa, A549, H1299, HCT116, Mouse liver.
Predicted reactivity: Pig (83%), Rhesus Monkey (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes lipase A, the lysosomal acid lipase (also known as cholesterol ester hydrolase). This enzyme functions in the lysosome to catalyze the hydrolysis of cholesteryl esters and triglycerides. Mutations in this gene can result in Wolman disease and cholesteryl ester storage disease. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acid cholesteryl ester hydrolase; cholesterol ester hydrolase; cholesteryl esterase; diacylglycerol lipase; lipase A; lipase A, lysosomal acid, cholesterol esterase; lysosomal acid lipase; lysosomal acid lipase 1; lysosomal acid lipase A; RP11-341B24.1; sterol esterase; triacylglycerol ester hydrolase; triacylglycerol lipase; unnamed protein product
Gene Aliases: CESD; LAL; Lip-1; Lip1
UniProt ID: (Human) P38571, (Mouse) Q9Z0M5
Entrez Gene ID: (Human) 3988, (Mouse) 16889
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support