Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Recommended positive controls: 293T, A431, Jurkat, Raji.
Predicted reactivity: Mouse (97%), Rat (98%), Zebrafish (85%), Xenopus laevis (88%), Chicken (94%), Rhesus Monkey (100%), Bovine (96%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by CRSP6 is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e. g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activator-recruited cofactor 77 kDa component; ARC77; cofactor required for Sp1 transcriptional activation, subunit 6, 77kDa; CRSP complex subunit 6; epididymis secretory sperm binding protein; thyroid hormone receptor-associated protein complex 80 kDa component; transcriptional coactivator CRSP77; unnamed protein product; vitamin D3 receptor-interacting protein complex 80 kDa component
Gene Aliases: CRSP6; CRSP77; DRIP80; SRB4; TRAP80
UniProt ID: (Human) Q9NVC6
Entrez Gene ID: (Human) 9440
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support