Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Peptide sequence: PPAFGLFDDA AAAAAALGLA PPAARGLLTP PASPLELLEA KPKRGRRSWP
Sequence homology: Guinea Pig: 93%; Human: 100%; Mouse: 93%; Pig: 100%; Rat: 93%; Zebrafish: 79%
KLF2 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 3 C2H2-type zinc fingers. KLF2 binds to the CACCC box in the beta-globin gene promoter and activates transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kruppel like factor 2; Kruppel-like factor 2 (lung); Kruppel-like factor LKLF; lung krueppel-like factor; lung Kruppel-like factor; lung Kruppel-like zinc finger transcription factor; zinc finger protein; LKLF
Gene Aliases: LKLF
UniProt ID: (Human) Q9Y5W3
Entrez Gene ID: (Human) 10365
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support