Search
Search
NeoBiotechnologies
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive Control:Muscle or sarcoma.
Cellular Location: Cytoplasm.
The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACTA1; ACTA2; ACTC; ACTG2; actin, alpha 2, smooth muscle, aorta; actin, gamma 2, smooth muscle, enteric; actin-like protein; alpha-actin; alpha-actin-3; alpha-cardiac actin; alpha-sma; cell growth-inhibiting gene 46 protein; cytoskeletal gamma-actin; epididymis luminal protein 176; gamma-actin; GIG46; nemaline myopathy type 3; RP5-1068B5.2; unnamed protein product
Gene Aliases: ACT; ACTA; ACTA3; ACTC; ACTE; ACTG; ACTL3; ACTSA; ACTSG; ASD5; ASMA; CFTD; CFTD1; CFTDM; CMD1R; CMH11; CMYO2A; CMYO2B; CMYO2C; CMYP2A; CMYP2B; CMYP2C; DFNA20; DFNA26; HEL-176; LVNC4; MMIHS5; MPFD; NEM1; NEM2; NEM3; SHPM; SMDYS; VSCM; VSCM1
UniProt ID: (Human) P68133, (Human) P62736, (Human) P68032, (Human) P63261, (Human) P63267
Entrez Gene ID: (Human) 58, (Human) 59, (Human) 70, (Human) 71, (Human) 72
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support