Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (64%), Rat (64%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84647. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
LYSMD1 (LysM Domain Containing 1) is a Protein Coding gene. Diseases associated with LYSMD1 include 3-Methylglutaconic Aciduria, Type Vii. An important paralog of this gene is LYSMD2. [GeneCards]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LysM, putative peptidoglycan-binding, domain containing 1; RP11-68I18.5; unnamed protein product
Gene Aliases: SB145
UniProt ID: (Human) Q96S90
Entrez Gene ID: (Human) 388695
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support