Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
It is recommended that the protein be aliquoted for optimal storage.
It is recommended that sterile water (650 µL) be added to the vial to prepare a stock solution of 0.77 mg/mL.
Concentration is measured by UV.
CD85j, also known as a member of the leukocyte immunoglobulin-like receptor (LIR) family, is a single transmembrane glycoprotein with a long cytoplasmic domain containing four immunoreceptor tyrosine-based inhibitory motifs (ITIMs). These motifs signal through interactions with SHP-1, transducing negative signals that inhibit immune response stimulation. CD85j is expressed on myeloid cells such as monocytes and dendritic cells, as well as on subsets of lymphoid cells including NK, T, and B cells. The expression of CD85j is correlated with leukemias such as acute lymphoblastic leukemia (ALL) and chronic lymphocytic leukemia (CLL). On CD8+ cells, CD85j expression is associated with effector cell function and plays a significant role in viral infections, including HIV, Epstein-Barr virus, and cytomegalovirus (CMV). CD85j binds to MHC class I molecules, including HLA-G, A, F, B27, E, and F, on antigen-presenting cells. The receptor helps control inflammatory responses and cytotoxicity, focusing the immune response and limiting autoreactivity. CD85j belongs to the subfamily B class of LIR receptors, which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and multiple transcript variants encoding different isoforms have been identified for this gene. Its gene is located in a cluster at chromosomal region 19q13.4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD85 antigen-like family member J; Ig-like transcript 2; ILT2a; allelic form of ILT2; ILT2b; alternatively spliced form of ILT2; ILT2c; alternatively spliced form of ILT2; immunoglobulin heavy chain variable region; leucocyte Ig-like receptor B1; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1; LIR-1; monocyte/macrophage immunoglobulin-like receptor 7; myeloid inhibitory receptor 7; unnamed protein product
Gene Aliases: CD85J; ILT-2; ILT2; LIR-1; LIR1; MIR-7; MIR7; PIR-B; PIRB
UniProt ID: (Human) Q8NHL6
Entrez Gene ID: (Human) 10859
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support