Labtainer™ BioProcess Container (BPC), 250mL with 2 Ports, Luer Lock, 2D BPC
El producto real puede variar

Labtainer™ BioProcess Container (BPC), 250mL with 2 Ports, Luer Lock, 2D BPC

Have Questions?
Número de catálogo SH3065713
también denominado SH30657.13
Precio (MXN)
-
The 250 mL Thermo Scientific™ Labtainer™ BPC, 2D system is composed of CX5-14 single-web, multi-layer film, and includes 2 edge ports (male and female luer lock connections).

Tubing style: no tubing

Features:
• Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
• All Labtainers are constructed in an ISO 7 cleanroom under cGMP conditions.

Applications of the Thermo Scientific Labtainer BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors.
• Buffer, culture media, and process liquid preparation and storage.
• Chromatography feed and fraction collection.
• Harvest, storage and transport of bulk drug product and bulk drug precursors.

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library – with over 20,000 unique components that include:
Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
Filters—used for bioburden reduction; sterilization grade filters and vent filters.
Tubing—thermoplastic elastomers, platinum-cured silicone.
Sample ports—septum-style and needle-free ports.
Clamps—sliders, pinch, and crimp bands.

Related products:
Labtainer™ BioProcess Container (BPC), 50 mL with 2 Ports, Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 100 mL with 2 Ports, Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 250 mL with 2 Ports, Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 500 mL with 2 Ports, Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 1 L with 2 Ports, Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 2 L with 2 Ports, Luer Lock, 2D BPC

Labtainer™ BioProcess Container (BPC), 50 mL with 2 Ports, Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 100 mL with 2 Ports, Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 250 mL with 2 Ports, Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 500 mL with 2 Ports, Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 1 L with 2 Ports, Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 2 L with 2 Ports, Luer Lock, 2D BPC

Labtainer™ BioProcess Container (BPC), 50 mL with 2 Ports, Luer Lock and MPC Insert, 2D BPC
Labtainer™ BioProcess Container (BPC), 100 mL with 2 Ports, Luer Lock and MPC Insert, 2D BPC
Labtainer™ BioProcess Container (BPC), 250 mL with 2 Ports, Luer Lock and MPC Insert, 2D BPC
Labtainer™ BioProcess Container (BPC), 500 mL with 2 Ports, Luer Lock and MPC Insert, 2D BPC
Labtainer™ BioProcess Container (BPC), 1 L with 2 Ports, Luer Lock and MPC Insert, 2D BPC
Labtainer™ BioProcess Container (BPC), 2 L with 2 Ports, Luer Lock and MPC Insert, 2D BPC

Labtainer™ BioProcess Container (BPC), 5 L with 3 Ports, MPC Insert and Injection Port, 2D BPC
Labtainer™ BioProcess Container (BPC), 10 L with 3 Ports, MPC Insert and Injection Port, 2D BPC
Labtainer™ BioProcess Container (BPC), 20 L with 3 Ports, MPC Insert and Injection Port, 2D BPC
Labtainer™ BioProcess Container (BPC), 50 L with 3 Ports, MPC Insert and Injection Port, 2D BPC
Labtainer™ BioProcess Container (BPC), 2 L with 3 Ports, MPC Insert and Injection Port, 2D BPC

Labtainer™ BioProcess Container (BPC), 5 L with 3 Ports, MPC Insert, MPC Body, and Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 10 L with 3 Ports, MPC Insert, MPC Body, and Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 20 L with 3 Ports, MPC Insert, MPC Body, and Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 50 Lwith 3 Ports, MPC Insert, MPC Body, and Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 2 L with 3 Ports, MPC Insert, MPC Body, and Luer Lock, 2D BPC

Labtainer™ BioProcess Container (BPC), 5 L with 3 Ports, Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 10 L with 3 Ports, Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 20 L with 3 Ports, Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 50 L with 3 Ports, Luer Lock, 2D BPC
Labtainer™ BioProcess Container (BPC), 2 L with 3 Ports, Luer Lock, 2D BPC

Labtainer™ BioProcess Container (BPC), 5 L with 3 Ports, MPC Insert, 2D BPC
Labtainer™ BioProcess Container (BPC), 10 L with 3 Ports, MPC Insert, 2D BPC
Labtainer™ BioProcess Container (BPC), 20 L with 3 Ports, MPC Insert, 2D BPC
Labtainer™ BioProcess Container (BPC), 50 L with 3 Ports, MPC Insert, 2D BPC
Labtainer™ BioProcess Container (BPC), 2 L with 3 Ports, MPC Insert, 2D BPC

Labtainer™ BioProcess Container (BPC), 50L with 3 Ports, 2D BPC
Labtainer™ BioProcess Container (BPC), 100L with 3 Ports, 2D BPC
Labtainer™ BioProcess Container (BPC), 200L with 3 Ports, 2D BPC
Especificaciones
Unit Size250 mL