Labtainer™ BioProcess Container (BPC), 20L with 3 Ports, MPC Insert and Injection Port, 2D BPC
Thermo Scientific™

Labtainer™ BioProcess Container (BPC), 20L with 3 Ports, MPC Insert and Injection Port, 2D BPC

The 20 L Thermo Scientific™ Labtainer™ BPC, 2D system is composed of CX5-14 single web, multi-layer plastic film and includesRead more
Have Questions?
Catalog NumberQuantity
SH307090320 L, each
Catalog number SH3070903
Price (USD)
-
Quantity:
20 L, each
The 20 L Thermo Scientific™ Labtainer™ BPC, 2D system is composed of CX5-14 single web, multi-layer plastic film and includes 3 top ports (2 with MPC inserts, 1 injection port).

Features:
• Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
• All Labtainers are constructed in an ISO 7 cleanroom under cGMP conditions.

Applications of the Thermo Scientific Labtainer BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors.
• Buffer, culture media, and process liquid preparation and storage.
• Chromatography feed and fraction collection.
• Harvest, storage and transport of bulk drug product and bulk drug precursors.

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library – with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
• Filters—used for bioburden reduction; sterilization grade filters and vent filters.
• Tubing—thermoplastic elastomers, platinum-cured silicone.
• Sample ports—septum-style and needle-free ports.
• Clamps—sliders, pinch, and crimp bands.

Specifications
Dimensions25.8 x 17 in. (65.5 x 43.2 cm)
Outer PackagingPackages placed in strong cardboard cartons with product and lot identification
Product LineLabtainer
Protective Inner PackagingPacked and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity20 L, each
Sterilization MethodGamma Irradiated
Tubing Line 1Line 1. MPC insert, polycarbonate, No Tubing
Tubing Line 2Line 2. MPC insert, polycarbonate,No Tubing
Tubing Line 3Line 3. Injection Port, No Tubing
Capacity (Metric)20 L
Film DescriptionCX5-14
Port DetailEdge Ports: 2 x 0.38 in. 1 x 0.25 in.
Unit Size20 L