Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MDPNCSCTT GVSCACTGSC KCKECKCTSC KKSCCSCCPV GCAKCAHGCV CKGTLENCSC CA (1-61 aa encoded by BC028280)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: metallothionein 1K; metallothionein 1Y; Metallothionein-1M; Metallothionein-IM; MT-1M; MT-IM
Gene Aliases: MT-1M; MT-IM; MT1; MT1K; MT1M
UniProt ID: (Human) Q8N339
Entrez Gene ID: (Human) 4499
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support