Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SWKEPCQVQ SCQEEMISLG DGDIRTMCPL LSMKPGEYSY FSPRTMSMWA GPDHWRFRPR RKQDAPSQSE NKKKSTKKDF EIDFEDDIDF DVYFRKTKAA TILTKSTLEN QNWRATTLPT DFNYNVDTLV QLHLKPGTRL LKMAQGHRAE TEHYEEIEDY DYNNPNDTSN FCPGLQAADS DDEDLDDLFV GPVGNSDLSP YPCHPPKTAQ QNGDTPEAQG LDITTYGESN LVAEPQKVNK IEIHYAKTAK KMDMKKLKQS MWSLLTALSG KEADAEANHR EAGKEAALAE VADEKMLSGL TKDLQRSLPP VMAQNLSIPL AFACLLHLAN EKNLKLEGTE DLSDVLVRQG D (392-741 aa encoded by BC024211)
This gene encodes a member of the barr gene family and a regulatory subunit of the condensin complex. This complex is required for the conversion of interphase chromatin into condensed chromosomes. The protein encoded by this gene is associated with mitotic chromosomes, except during the early phase of chromosome condensation. During interphase, the protein has a distinct punctate nucleolar localization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: barren homolog 1; Barren homolog protein 1; Chromosome-associated protein H; Condensin complex subunit 2; HCAP-H; mCAP-H; Non-SMC condensin I complex subunit H; unnamed protein product; XCAP-H homolog
Gene Aliases: A730011O11Rik; BRRN; BRRN1; CAP-H; CAPH; HCAP-H; KIAA0074; MCPH23; mKIAA0074; NCAPH; NCAPH1
UniProt ID: (Human) Q15003, (Mouse) Q8C156
Entrez Gene ID: (Human) 23397, (Mouse) 215387
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support