Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MSVQVAAPG SAGLGPERLS PEELVRQTRQ VVQGLEALRA EHHGLAGHLA EALAGQGPAA GLEMLEEKQQ VVSHSLEAIE LGLGEAQVLL ALSAHVGALE AEKQRLRSQA RRLAQENVWL REELEETQRR LRASEESVAQ LEEEKRHLEF LGQLRQYDPP AESQVPRAGR GGGCWALHRA PQSPRPSLES HSPQDPP (1-196 aa encoded by BC025318)
KLC3 encodes a member of the kinesin light chain gene family. Kinesins are molecular motors involved in the transport of cargo along microtubules, and are composed of two kinesin heavy chain (KHC) and two kinesin light chain (KLC) molecules. KLCs are thought to typically be involved in binding cargo and regulating kinesin activity. In the rat, a protein similar to this gene product is expressed in post-meiotic spermatids, where it associates with structural components of sperm tails and mitochondria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kinesin light chain 2; Kinesin light chain 3; Kinesin light chain KLCt; KLC2-like; unnamed protein product
Gene Aliases: BC017147; KLC2; KLC2L; KLC3; KLCt; KNS2B
UniProt ID: (Human) Q6P597, (Mouse) Q91W40, (Rat) Q68G30
Entrez Gene ID: (Human) 147700, (Mouse) 232943, (Rat) 171549
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support