Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: EQARAELLS LTQMEREKIV WEFEQLYHSL KEHEYRLLAR LEELDLAIYN SINGAITQFS CNISHLSSLI AQLEEKQQQP TRELLQDIGD TLSRAERIRI PEPWITPPDL QEKIHIFAQK CLFLTESLKQ FTEKMQSDME KIQELREAQL YSVDVTLDPD TAYPSLILSD NLRQVRYSYL QQDLPDNPER FNLFPCVLGS PCFIAGRHYW EVEVGDKAKW TIGVCEDSVC RKGGVTSAPQ NGFWAVSLWY GKEYWALTSP MTALPLRTPL QRVGIFLDYD AGEVSFYNVT ERCHTFTFSH ATFCGPVRPY FSLSYSGGKS AAPLIICPMS GIDGFSGHVG NHGHSMETSP (165-514 aa encoded by BC013580)
This gene encodes a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to the nuclear matrix. It interacts with the enhancer of polycomb protein and represses gene transcription. It is also thought to be involved in the differentiation of male germ cells. Fusion of the N-terminus of this protein with the truncated C-terminus of the RET gene product has been shown to result in production of the ret transforming protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ret finger protein; ret finger protein 1; RFP transforming protein; RING finger protein 76; RING-type E3 ubiquitin transferase TRIM27; tripartite motif protein 27; tripartite motif protein TRIM27; Tripartite motif-containing protein 27; unnamed protein product; Zinc finger protein RFP
Gene Aliases: AW538890; Gm19403; RFP; Rfp1; RNF76; TRIM27
UniProt ID: (Human) P14373, (Mouse) Q62158
Entrez Gene ID: (Human) 5987, (Mouse) 19720, (Rat) 291171
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support