Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MPCIQAQYG TPAPSPGPRD HLASDPLTPE FIKPTMDLAS PEAAPAAPTA LPSFSTFMDG YTGEFDTFLY QLPGTVQPCS SASSSASSTS SSSATSPASA SFKFEDFQVY GCYPGPLSGP VDEALSSSGS DYYGSPCSAP SPSTPSFQPP QLSPWDGSFG HFSPSQTYEG LRAWTEQLPK ASGPPQPPAF FSFSPPTGPS PSLAQSPLKL FPSQATHQLG EGESYSMPTA FPGLAPTSPH LEGSGILDTP VTSTKARSGA PGGSEGRCAV (1-269 aa encoded by BC016147)
NR4A1 is a member of the steroid-thyroid hormone-retinoid receptor superfamily. NGFI-B-alpha is expressed in brain, adrenal, thyroid, liver, testis, ovary, thymus, muscle, lung and prostate. Its expression is induced in response to various stress stimuli and growth factors. The encoded protein acts as a nuclear transcription factor involved in mediating apoptotic signaling in thymocytes and tumor cells, as well as having a role in signaling in the hypothalamic-pituitary axis. Translocation of the protein from the nucleus to mitochondria induces apoptosis. Three transcript variants encoding two distinct isoforms have been identified for this NR4A1. Variant (3), also known as TRC beta, contains multiple differences compared to variant 1. The resulting isoform b contains a shorter and distinct C-terminus compared to isoform a. RXR has been shown to be a partner for NR4A1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Early response protein NAK1; growth factor-inducible nuclear protein N10; hormone receptor; MGC9485; NR4A1; Nuclear hormone receptor NUR/77; Nuclear protein N10; Nuclear receptor subfamily 4 group A member 1; Nuclear receptor subfamily 4immunitygroup A member 1; Nur77; Orphan nuclear receptor HMR; Orphan nuclear receptor TR3; Orphan receptor tr3; ST-59; steroid receptor TR3; Testicular receptor 3; TR3 orphan receptor; unnamed protein product
Gene Aliases: Gfrp; GFRP1; Hbr-1; Hbr1; HMR; N10; NAK-1; NAK1; NGFI-B; NGFIB; NP10; NR4A1; NUR77; TIS1; TR3
UniProt ID: (Human) P22736, (Mouse) P12813
Entrez Gene ID: (Human) 3164, (Mouse) 15370
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support