Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LTDEEVEMT RMAVQFELED LNLRPDPEPL LTEMIHEAAY RENTVGLHRF CPTENVAKIN REVLHSYLRN YYTPDRMVLA GVGVEHEHLV DCARKYLLGV QPAWGSAEAV DIDRSVAQYT GGIAKLERDM SNVSLGPTPI PELTHIMVGL ESCSFLEEDF IPFAVLNMMM GGGGSFSAGG PGKGMFSRLY LNVLNRHHWM YNATSYHHSY EDTGLLCIHA SADPRQVREM VEIITKEFIL MGGTVDTVEL ERAKTQLTSM LMMNLESRPV IFEDVGRQVL ATRSRKLPHE LCTLIRNVKP EDVKRVASKM LRGKPAVAAL GDLTDLPTYE HIQTALSSKD GRLPRTYRLF R (176-525 aa encoded by BC132724)
Substrate recognition and binding subunit of the essential mitochondrial processing protease (MPP), which is required for maturation of the majority of mitochondrial precursor proteins. Most MPP cleavage sites follow an arginine at position -2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-MPP; cerebellar ataxia 1 (autosomal recessive); Cerebellar ataxia-1; Inactive zinc metalloprotease alpha; inositol polyphosphate-5-phosphatase, 72 kD; KIAA0123; mitochondrial matrix processing protease, alpha subunit; Mitochondrial-processing peptidase subunit alpha; P-55; peptidase, mitochondrial processing alpha subunit; unnamed protein product
Gene Aliases: Alpha-MPP; CLA1; CPD3; INPP5E; KIAA0123; MAS2; MPPA; P-55; PMPCA; SCAR2
UniProt ID: (Human) E7ET61
Entrez Gene ID: (Human) 23203
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support